Thermo Scientific Chemicals

4-Bromo-2-methylpyridine, 97%, Thermo Scientific Chemicals

Catalog number: H27884.MD
250 mg, Each
Thermo Scientific Chemicals

4-Bromo-2-methylpyridine, 97%, Thermo Scientific Chemicals

Catalog number: H27884.MD
250 mg, Each
Quantity
Catalog number: H27884.MD
also known as H27884-MD
Price (USD)

Chemical Identifiers

CAS
22282-99-1
IUPAC Name
4-bromo-2-methylpyridine
Molecular Formula
C6H6BrN
InChI Key
JFBMFWHEXBLFCR-UHFFFAOYSA-N
SMILES
CC1=CC(Br)=CC=N1
Assay (GC)
≥96.0%
Refractive Index
1.5545-1.5595 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless to yellow to brown

Description

4-Bromo-2-methylpyridine is used as a starting material in the preparation of crown-ester-bipyridines and viologens through sodium or nickel reductive coupling, side chain oxidation and esterification. Further, it serves as an important raw material and intermediate in organic synthesis, pharmaceuticals, agrochemicals and in dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-methylpyridine is used as a starting material in the preparation of crown-ester-bipyridines and viologens through sodium or nickel reductive coupling, side chain oxidation and esterification. Further, it serves as an important raw material and intermediate in organic synthesis, pharmaceuticals, agrochemicals and in dyes.

Solubility
Miscible with dichloromethane. Immiscible with water.

Notes
Light and moisture sensitive. Incompatible with strong oxidizing agents, reducing agents, strong acids and bases. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text