Furan-2,5-dicarboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Furan-2,5-dicarboxylic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H28718.06
also known as H28718-06
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS3238-40-2
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White to pale cream
Assay (Silylated GC)≥97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only