Thermo Scientific Chemicals

4-Chloro-6,7-dihydro-5H-cyclopenta[4,5]thieno[2,3-d]pyrimidine, 96%, Thermo Scientific Chemicals

Catalog number: H32208.06
5 g, Each
Thermo Scientific Chemicals

4-Chloro-6,7-dihydro-5H-cyclopenta[4,5]thieno[2,3-d]pyrimidine, 96%, Thermo Scientific Chemicals

Catalog number: H32208.06
5 g, Each
Quantity
Catalog number: H32208.06
also known as H32208-06
Price (USD)

Chemical Identifiers

CAS
300816-22-2
IUPAC Name
12-chloro-7-thia-9,11-diazatricyclo[6.4.0.0²,⁶]dodeca-1(12),2(6),8,10-tetraene
Molecular Formula
C9H7ClN2S
InChI Key
JKXJMJPJOYMQSK-UHFFFAOYSA-N
SMILES
ClC1=C2C3=C(CCC3)SC2=NC=N1
Appearance (Color)
Pale brown
Form
Powder
Melting Point (clear melt)
103.0-112.0?C
Assay (GC)
≥95.0%

Description

Used in the synthesis of some novel thieno [2, 3-d] pyrimidines and their antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of some novel thieno [2, 3-d] pyrimidines and their antibacterial activity.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text