Thermo Scientific Chemicals

3-Bromo-2-fluorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: H32228.06
5 g, Each
Thermo Scientific Chemicals

3-Bromo-2-fluorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: H32228.06
5 g, Each
Quantity
Catalog number: H32228.06
also known as H32228-06
Price (USD)

Chemical Identifiers

CAS
149947-16-0
IUPAC Name
1-bromo-3-(bromomethyl)-2-fluorobenzene
Molecular Formula
C7H5Br2F
InChI Key
MGVHFTWDCYIBDO-UHFFFAOYSA-N
SMILES
FC1=C(CBr)C=CC=C1Br
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Melting Point (clear melt)
32.0-41.0?C
Assay (GC)
≥96.0%
Appearance (Color)
Pale orange or pale cream

Description

Used to obtain the target inhibitors. A new quasi-one-dimensional molecular solid based on Ni (mnt) 2 anion: Crystal structure and spin-gap transition 4-Bromo-2-fluorobenzyl bromide acts as stasrting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to obtain the target inhibitors. A new quasi-one-dimensional molecular solid based on Ni (mnt) 2 anion: Crystal structure and spin-gap transition 4-Bromo-2-fluorobenzyl bromide acts as stasrting material.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text