Thermo Scientific Chemicals

2-Chloro-4,6-diphenyl-1,3,5-triazine, 97%, Thermo Scientific Chemicals

Catalog number: H33175.06
5 g, Each
Thermo Scientific Chemicals

2-Chloro-4,6-diphenyl-1,3,5-triazine, 97%, Thermo Scientific Chemicals

Catalog number: H33175.06
5 g, Each
Quantity
Catalog number: H33175.06
also known as H33175-06
Price (USD)

Chemical Identifiers

CAS
3842-55-5
IUPAC Name
2-chloro-4,6-diphenyl-1,3,5-triazine
Molecular Formula
C15H10ClN3
InChI Key
DDGPPAMADXTGTN-UHFFFAOYSA-N
SMILES
ClC1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to cream
Form
Powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
134.0-143.0?C

Description

A rational synthesis of kyaphenine (2:4:6-triphenyl-1:3:5-triazine) has been effected by reaction of 2-chloro-4:6-diphenyl-1:3:5-triazine with phenylmagnesium bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A rational synthesis of kyaphenine (2:4:6-triphenyl-1:3:5-triazine) has been effected by reaction of 2-chloro-4:6-diphenyl-1:3:5-triazine with phenylmagnesium bromide.

Solubility
Soluble in toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text