Thermo Scientific Chemicals

(2-Bromophenylethynyl)trimethylsilane, 98%, Thermo Scientific Chemicals

Catalog number: H53402.14
25 g, Each
Thermo Scientific Chemicals

(2-Bromophenylethynyl)trimethylsilane, 98%, Thermo Scientific Chemicals

Catalog number: H53402.14
25 g, Each
Quantity
Catalog number: H53402.14
also known as H53402-14
Price (USD)

Description

(2-Bromophenylethynyl)trimethylsilane finds wide range application in the medicine and electronic industry. Reaction of (2-bromophenylethynyl)trimethylsilane 1 with trimethyl borate yielded the alkynearylboronic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(2-Bromophenylethynyl)trimethylsilane finds wide range application in the medicine and electronic industry. Reaction of (2-bromophenylethynyl)trimethylsilane 1 with trimethyl borate yielded the alkynearylboronic acid.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text