4-Bromo-N-methylbenzylamine, 97%, Thermo Scientific Chemicals
4-Bromo-N-methylbenzylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-N-methylbenzylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H54506.03
also known as H54506-03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS699-03-6
IUPAC Name[(4-bromophenyl)methyl](methyl)amine
Molecular FormulaC8H10BrN
InChI KeyURFJXIULELMVHV-UHFFFAOYSA-N
SMILESCNCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only