Thermo Scientific Chemicals

2-Cyanopyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog number: H54689.MD
250 mg, Each
Thermo Scientific Chemicals

2-Cyanopyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog number: H54689.MD
250 mg, Each
Quantity
Catalog number: H54689.MD
also known as H54689-MD
Price (USD)
Have Questions?

Chemical Identifiers

CAS
741709-63-7
IUPAC Name
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carbonitrile
Molecular Formula
C12H15BN2O2
InChI Key
IXTBQKLZPOYJFJ-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CN=C(C=C1)C#N
Appearance (Color)
White
Proton NMR
Conforms to structure
Form
Powder

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is used in suzuki reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is used in suzuki reactions.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents, air, heat. Refrigerated (2-4°C).
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text