Thermo Scientific Chemicals

Bis(pentafluorophenyl) carbonate, 98+%, Thermo Scientific Chemicals

Catalog number: H54880.14
25 g, Each
Thermo Scientific Chemicals

Bis(pentafluorophenyl) carbonate, 98+%, Thermo Scientific Chemicals

Catalog number: H54880.14
25 g, Each
Quantity
Catalog number: H54880.14
also known as H54880-14
Price (USD)

Chemical Identifiers

CAS
59483-84-0
Assay (GC)
>98.0%

Description

Bis(pentafluorophenyl) carbonate is a reagent used as a carbonyl equivalent and for various coupling reactions, especially for azapeptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(pentafluorophenyl) carbonate is a reagent used as a carbonyl equivalent and for various coupling reactions, especially for azapeptides.

Solubility
Soluble in chloroform, ethyl acetate and methylene chloride.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text