Thermo Scientific Chemicals

Methyl 4-vinylbenzoate, 96%, Thermo Scientific Chemicals

Catalog number: H60057.06
5 g, Each
Thermo Scientific Chemicals

Methyl 4-vinylbenzoate, 96%, Thermo Scientific Chemicals

Catalog number: H60057.06
5 g, Each
Quantity
Catalog number: H60057.06
also known as H60057-06
Price (USD)

Chemical Identifiers

CAS
1076-96-6
IUPAC Name
methyl 4-ethenylbenzoate
Molecular Formula
C10H10O2
InChI Key
NUMHUJZXKZKUBN-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(C=C)C=C1
Assay (HPLC)
>96.0%

Description

Methyl 4-vinylbenzoate is used as a fluid component for enhancing ink stability in printing and also in preparing vinyl substituted beta-diketones. Further, it is used as an organic intermediate for pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-vinylbenzoate is used as a fluid component for enhancing ink stability in printing and also in preparing vinyl substituted beta-diketones. Further, it is used as an organic intermediate for pharmaceuticals.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text