Thermo Scientific Chemicals

3,5-Dimethoxybenzylamine, 98%, Thermo Scientific Chemicals

Catalog number: H60503.06
5 g, Each
Thermo Scientific Chemicals

3,5-Dimethoxybenzylamine, 98%, Thermo Scientific Chemicals

Catalog number: H60503.06
5 g, Each
Quantity
Catalog number: H60503.06
also known as H60503-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
34967-24-3
IUPAC Name
1-(3,5-dimethoxyphenyl)methanamine
Molecular Formula
C9H13NO2
InChI Key
YGZJTYCCONJJGZ-UHFFFAOYSA-N
SMILES
COC1=CC(OC)=CC(CN)=C1
Melting Point
33-39°C
Appearance (Color)
White to pale yellow (Solid) or colorless to pale yellow (Liquid)
Form
Crystals or powder or crystalline powder or fused solid or chunks
Assay (GC)
≥97.5%

Description

3,5-Dimethoxybenzylamine was used in the preparation of trisammonium tris(hexafluoro phosphate) salt.  It was also used in the synthesis of well defined, homogeneous [n]rotaxanes (n up to 11) by a template-directed thermodynamic clipping approach.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethoxybenzylamine was used in the preparation of trisammonium tris(hexafluoro phosphate) salt.  It was also used in the synthesis of well defined, homogeneous [n]rotaxanes (n up to 11) by a template-directed thermodynamic clipping approach.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text