Thermo Scientific Chemicals

Chlortetracycline hydrochloride, Thermo Scientific Chemicals

Catalog number: J60095.14
25 g, Each
Thermo Scientific Chemicals

Chlortetracycline hydrochloride, Thermo Scientific Chemicals

Catalog number: J60095.14
25 g, Each
Quantity
Catalog number: J60095.14
also known as J60095-14
Price (USD)

Chemical Identifiers

CAS
64-72-2
IUPAC Name
hydrogen (2Z,4S,4aS,5aS,6S,12aS)-2-[amino(hydroxy)methylidene]-7-chloro-4-(dimethylamino)-6,10,11,12a-tetrahydroxy-6-methyl-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
Molecular Formula
C22H24Cl2N2O8
InChI Key
RRXOSDMYCQSQHB-MRYPCDLHSA-N
SMILES
[H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)C4=C(O)C=CC(Cl)=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)C(=C(N)O)C1=O
Form
Powder
Loss on Drying
2.0% max.
Appearance (Color)
Yellow
Assay (unspecified)
Potency: 900?g/mg min.
pH
2.3 to 3.3 (1% w/v in water)

Description

Chlortetracycline was used in fluorescence assays to detect the Ca+2 influx required for the completion of acrosome reaction in human spermatozoa.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlortetracycline was used in fluorescence assays to detect the Ca+2 influx required for the completion of acrosome reaction in human spermatozoa.

Solubility
Soluble in water

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text