Hexestrol, 98+%, Thermo Scientific Chemicals
Hexestrol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hexestrol, 98+%, Thermo Scientific Chemicals

Nonsteroidal synthetic estrogen
Have Questions?
Quantity:
1 g
Catalog number J60889.03
also known as J60889-03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHexestrol
CAS84-16-2
Health Hazard 1H350
Health Hazard 2GHS H Statement
H350
May cause cancer.
Health Hazard 3P201-P202-P281-P308+P313-P501c
View more
Nonsteroidal synthetic estrogen

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only