Glycine, Electrophoresis Grade, 99+%
Thermo Scientific Chemicals
Glycine, Electrophoresis Grade, 99+%
Have Questions?
Quantity:
250 g
Catalog number J64365.30
also known as J64365-30
Price (USD)/ Each
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS56-40-6
IUPAC Name2-aminoacetic acid
Molecular FormulaC2H5NO2
InChI KeyDHMQDGOQFOQNFH-UHFFFAOYSA-N
SMILESNCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline powder
Assay from Suppliers CofA≥99.0%
Loss on Drying≤0.2%
Glycine is used as a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis. Glycine is also a component of Towbin?s transfer buffer for Western blots. Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine is used as a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis. Glycine is also a component of Towbin′s transfer buffer for Western blots. Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Solubility
Freely soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Tapio Haaparanta.; Jouni Uitto.; Erkki Ruoslahti.; Eva Engvall. A determination of submicromolar concentrations of glycine in periaqueductal gray matter microdialyzates using capillary zone electrophoresis with contactless conductivity detection. Electrophoresis. 2009, 30, (19), 3436-3441.