Thermo Scientific Chemicals

N,N-Dimethylformamide di-tert-butyl Acetal, tech. 90%, Thermo Scientific Chemicals

Catalog number: L00431.06
5 g, Each
Thermo Scientific Chemicals

N,N-Dimethylformamide di-tert-butyl Acetal, tech. 90%, Thermo Scientific Chemicals

Catalog number: L00431.06
5 g, Each
Quantity
Catalog number: L00431.06
also known as L00431-06
Price (USD)

Chemical Identifiers

CAS
36805-97-7
IUPAC Name
[bis(tert-butoxy)methyl]dimethylazanium
Molecular Formula
C11H26NO2
InChI Key
DBNQIOANXZVWIP-UHFFFAOYSA-O
SMILES
C[NH+](C)C(OC(C)(C)C)OC(C)(C)C
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥88.0%
Refractive Index
1.4060-1.4150 @ 20?C

Description

N,N-Dimethylformamide di-tert-butyl acetal is used as a reagent in the synthesis of (S)-2-(1-tert-butoxycarbonyl-2-{4-[2-(5-methyl-2-phenyloxazol-4-yl)ethoxy]phenyl}ethylamino)benzoic acid methyl ester, 3-O-tert-butylmorphine, tert-butylesters of pyrrole and indolecarboxylic acids. Further, it is used in the preparation of indolizines by intermolecular cyclization of picolinium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylformamide di-tert-butyl acetal is used as a reagent in the synthesis of (S)-2-(1-tert-butoxycarbonyl-2-{4-[2-(5-methyl-2-phenyloxazol-4-yl)ethoxy]phenyl}ethylamino)benzoic acid methyl ester, 3-O-tert-butylmorphine, tert-butylesters of pyrrole and indolecarboxylic acids. Further, it is used in the preparation of indolizines by intermolecular cyclization of picolinium salts.

Solubility
Miscible with toluene and benzene.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text