Thermo Scientific Chemicals

9-Octadecynoic acid, 98%, Thermo Scientific Chemicals

Catalog number: L01294.03
1 g, Each
Thermo Scientific Chemicals

9-Octadecynoic acid, 98%, Thermo Scientific Chemicals

Catalog number: L01294.03
1 g, Each
Quantity
Catalog number: L01294.03
also known as L01294-03
Price (USD)

Chemical Identifiers

CAS
506-24-1
IUPAC Name
octadec-9-ynoic acid
Molecular Formula
C18H32O2
InChI Key
RGTIBVZDHOMOKC-UHFFFAOYSA-N
SMILES
CCCCCCCCC#CCCCCCCCC(O)=O
Appearance (Color)
White to cream to yellow to pale brown
Form
Crystals or powder or crystalline powder or lumps or waxy solid
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

9-Octadecynoic acid, is used as an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Octadecynoic acid, is used as an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text