Thermo Scientific Chemicals

Vinyltrimethylsilane, 97%, Thermo Scientific Chemicals

Catalog number: L02498.18
50 g, Each
Thermo Scientific Chemicals

Vinyltrimethylsilane, 97%, Thermo Scientific Chemicals

Catalog number: L02498.18
50 g, Each
Quantity
Catalog number: L02498.18
also known as L02498-18
Price (USD)

Chemical Identifiers

CAS
754-05-2
IUPAC Name
ethenyltrimethylsilane
Molecular Formula
C5H12Si
InChI Key
GCSJLQSCSDMKTP-UHFFFAOYSA-N
SMILES
C[Si](C)(C)C=C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
>96.0%
Refractive Index
1.3885-1.3935 @ 20?C
Solution Test
Clear (10% in diethyl ether)

Description

Vinyltrimethylsilane is used in semiconductor processing. It is involved in the Preparation of silyl-ethers by Rh(I) catalysis. Further, it is used in the preparation of trimethylsilylethyl sulfoxides which in the presence of fluoride ion generate sulfenate nucleophiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinyltrimethylsilane is used in semiconductor processing. It is involved in the Preparation of silyl-ethers by Rh(I) catalysis. Further, it is used in the preparation of trimethylsilylethyl sulfoxides which in the presence of fluoride ion generate sulfenate nucleophiles.

Solubility
Miscible with tetrahydrofuran, diethyl ether, benzene and dichloromethane.

Notes
Store in a cool place. Hygroscopic. Incompatible with strong oxidizing agents, strong acids and strong alkalis (bases).
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text