You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Not web orderable
The product
L02947.06
:
exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride, 98%, Thermo Scientific Chemicals
is not orderable in your country at the moment. Please contact our
Technical Support
for possible alternatives.