Thermo Scientific Chemicals

Tetrahydrothiopyran, 98%, Thermo Scientific Chemicals

Catalog number: L03063.03
1 g, Each
Thermo Scientific Chemicals

Tetrahydrothiopyran, 98%, Thermo Scientific Chemicals

Catalog number: L03063.03
1 g, Each
Quantity
Catalog number: L03063.03
also known as L03063-03
Price (USD)

Chemical Identifiers

CAS
1613-51-0
IUPAC Name
thiane
Molecular Formula
C5H10S
InChI Key
YPWFISCTZQNZAU-UHFFFAOYSA-N
SMILES
C1CCSCC1
Form
Liquid
Refractive Index
1.5040-1.5080 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
>97.5%

Description

It is a useful 5 c synthon for the synthesis of 3-cyclopentenones. It is a organic sulfide, that has shown to have antioxidant activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful 5 c synthon for the synthesis of 3-cyclopentenones. It is a organic sulfide, that has shown to have antioxidant activity.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect against electrostatic charges.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text