4-Chloro-2,5-dimethoxyaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-2,5-dimethoxyaniline, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L04202.14
also known as L04202-14
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS6358-64-1
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)116.0-122.0?C
Appearance (Color)Pale brown to brown to pale grey to grey
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only