3-Chlorobenzo[b]thiophene-2-carbonyl chloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Chlorobenzo[b]thiophene-2-carbonyl chloride, 95%, Thermo Scientific Chemicals
Have Questions?
Quantity:
1 g
Catalog number L11745.03
also known as L11745-03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS21815-91-8
IUPAC Name3-chloro-1-benzothiophene-2-carbonyl chloride
Molecular FormulaC9H4Cl2OS
InChI KeyGWKSSMDJEWPKCM-UHFFFAOYSA-N
SMILESClC(=O)C1=C(Cl)C2=CC=CC=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to brown to yellow to pale yellow
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
Melting Point (clear melt)108.5-117.5?C
Assay (HPLC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only