Thermo Scientific Chemicals

Benzyl (R)-(-)-glycidyl ether, 98+%, Thermo Scientific Chemicals

Catalog number: L13980.06
5 g, Each
Thermo Scientific Chemicals

Benzyl (R)-(-)-glycidyl ether, 98+%, Thermo Scientific Chemicals

Catalog number: L13980.06
5 g, Each
Quantity
Catalog number: L13980.06
also known as L13980-06
Price (USD)

Chemical Identifiers

CAS
14618-80-5
IUPAC Name
(2R)-2-[(benzyloxy)methyl]oxirane
Molecular Formula
C10H12O2
InChI Key
QNYBOILAKBSWFG-JTQLQIEISA-N
SMILES
C(OCC1=CC=CC=C1)[C@H]1CO1
Appearance (Color)
Clear colorless
Refractive Index
1.5160-1.5200 @ 20?C
Form
Liquid
Optical Rotation
-5.2 ± 0.2? (c=5 in toluene)
Assay (GC)
≥98.0%

Description

Benzyl (R)-(-)-glycidyl ether is used in the preparation of the lactone fragment of compactin and mevinolin. Chiron in the preparation of syn-1,3-polyols, dideoxynucleosides, and a spiroacetal cyanohydrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl (R)-(-)-glycidyl ether is used in the preparation of the lactone fragment of compactin and mevinolin. Chiron in the preparation of syn-1,3-polyols, dideoxynucleosides, and a spiroacetal cyanohydrin.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text