Thermo Scientific Chemicals

2-Chloro-5-(chloromethyl)pyridine, 96%, Thermo Scientific Chemicals

Catalog number: L19284.06
5 g, Each
Thermo Scientific Chemicals

2-Chloro-5-(chloromethyl)pyridine, 96%, Thermo Scientific Chemicals

Catalog number: L19284.06
5 g, Each
Quantity
Catalog number: L19284.06
also known as L19284-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
70258-18-3
IUPAC Name
2-chloro-5-(chloromethyl)pyridine
Molecular Formula
C6H5Cl2N
InChI Key
SKCNYHLTRZIINA-UHFFFAOYSA-N
SMILES
ClCC1=CC=C(Cl)N=C1
Form
Crystals or powder or crystalline powder or fused/lumpy solid or turbid liquid as melt
Appearance (Color)
White to yellow to orange
Assay (GC)
≥95.0%

Description

2-Chloro-5-(chloromethyl)pyridine is used for the synthesis of various pharmaceutical compounds. It can also be used for the synthesis of new neonicotinoid compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5-(chloromethyl)pyridine is used for the synthesis of various pharmaceutical compounds. It can also be used for the synthesis of new neonicotinoid compounds.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text