Thermo Scientific Chemicals

Methallyltri-n-butyltin, 98%, Thermo Scientific Chemicals

Catalog number: L20186.03
1 g, Each
Thermo Scientific Chemicals

Methallyltri-n-butyltin, 98%, Thermo Scientific Chemicals

Catalog number: L20186.03
1 g, Each
Quantity
Catalog number: L20186.03
also known as L20186-03
Price (USD)

Chemical Identifiers

CAS
67883-62-9
IUPAC Name
tributyl(2-methylprop-2-en-1-yl)stannane
Molecular Formula
C16H34Sn
InChI Key
CYIUFVYZGOCGEV-UHFFFAOYSA-N
SMILES
CCCC[Sn](CCCC)(CCCC)CC(C)=C
Assay (GC)
≥97.5%
Refractive Index
1.4845-1.4885 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid
Identification (FTIR)
Conforms

Description

Used as a methallylating reagent. Also used as a sole curative. It is used as a primary and secondary intermediates in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a methallylating reagent. Also used as a sole curative. It is used as a primary and secondary intermediates in pharmaceuticals.

Solubility
Fully miscible with water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text