P5515 Hydraulic Pressure Comparator
P5515 Hydraulic Pressure Comparator
Fluke™
P5515 Hydraulic Pressure Comparator
Fluke P5515 Pneumatic Calibrator. A pressure comparator supplies the same precisely controlled pressure to both a reference gauge and device under test for calibration. The P5515 is uniquely qualified for pressure calibration workload over 10,000 psi.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
P5515-140MViton Seals
P5515-2700G-11 Gauge (2700G-G70M)
P5515-2700G-1-C
also known as P5515-2700G-1/C
1 Gauge Accredited
P5515-2700G-33 Gauges (2700G-G70M, 2700G-G20M, 2700G-BG7M)
P5515-2700G-3-C
also known as P5515-2700G-3/C
3 Gauge Accredited
P5515-140M-EPEthylene Propylene Seals
Catalog number P5515-140M
Price (ARS)
-
Includes:
Viton Seals
Ask our AI about this Product
The Fluke Calibration P5515 Pressure Comparator when combined with the 2700G Reference Pressure Gauges becomes an easy-to-use alternative to traditional deadweight testers that covers a wide range of workload. For convenience, the P5515 is bundled together with selected 2700G Reference Pressure Gauges for a complete bench top pressure calibration solution. These pressure calibrators provide the accuracy, reliability, and capability you need to calibrate dial gauges, digital test gauges and pressure transmitters.
A pressure comparator supplies the same precisely controlled pressure to both a reference gauge and device under test for calibration. The P5515 is uniquely qualified for pressure calibration workload over 10,000 psi.
• Generates pressures to 20,000 psi (1400 bar)
• Operational with a wide range of fluids
• Built-in hand pump for system priming and large volume applications
• High quality screw press for pressure generation and fine pressure control
• Acrylic reservoir for visibility of fluid level and quality
• Portable
• Can be used with the 2700G Reference Pressure Gauge to provide a complete calibration solution
Make it a pressure calibrator (P5515-2700G)
The Fluke Calibration P5515 Pressure Comparator when combined with the 2700G Reference Pressure Gauges becomes an easy-to-use alternative to traditional deadweight testers that covers a wide range of workload. For convenience, the P5515 is bundled together with selected 2700G Reference Pressure Gauges for a complete bench top pressure calibration solution. These pressure calibrators provide the accuracy, reliability, and capability you need to calibrate dial gauges, digital test gauges and pressure transmitters.
For example, the P5515-2700G-3 is a P5515 Pressure Comparator, bundled together with 2700G-G70M, 2700G-G20M, 2700G-BG7M. This calibrator has three pressure ranges corresponding to the three 2700G Reference Pressure Gauges in the bundle and covers a total pressure range of –12 psi to 10,000 psi (–80 kPa to 70 MPa).
These instruments are used for checking pressure measuring instruments against master test gauges, indicators or transducers. These hydraulic pressure comparators provide a quick and easy solution for checking pressure measuring instruments to 20,000 psi (1400 bar). Designed for operation with a wide range of fluids the 5515 include several features from our popular line of deadweight testers.
Specifications
DescriptionHydraulic Pressure Comparator
IncludesViton Seals
Pressure Range0 to 20 000 psi (140 MPa)
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link