Thermo Scientific™

Impeller Sleeve for Open-Top Mixing, for 200 L HyPerforma™ Single-Use Mixer BPC

Catalog number: SH30749.08
Thermo Scientific™

Impeller Sleeve for Open-Top Mixing, for 200 L HyPerforma™ Single-Use Mixer BPC

Catalog number: SH30749.08
Catalog Number
SH30749.08
Unit Size
Price (ARS)
Catalog NumberUnit SizePrice (ARS)
SH30749.08Contact Us ›
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Impeller Sleeve for Open-Top Mixing (200 L) supports mixing applications in bioproduction using Thermo Scientific Bioprocess Containers (BPCs) for Thermo Scientific HyPerforma Single-Use Mixer, with impeller-based mixing in open or closed designs.

Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability

Specifications

For Use With (Equipment)
HyPerforma 50 L Single-Use Mixer, DS 300 Single-Use Mixer
Product Type
Impeller Sleeve
Sterility
Sterile
Capacity (Metric)
200 L
Product Line
HyPerforma™

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text