Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Description | Transfer Assembly, 1 inch ID TPE, PendoTECH Pressure Sensor and AseptiQuik L Connectors |
Quantity | Each |
Tubing Style | TPE |
Connector Types | PendoTECH Pressure Sensor and AseptiQuik L Connectors |
Tubing Material | Pharmaceutical-grade thermoplastic elastomer |
Unit Size | 1 each |
Description | Transfer Assembly, 1 in. ID Silicone, PendoTECH Pressure Sensor and AseptiQuik L Connectors |
For Use With (Application) | Monoclonal antibody production, upstream and downstream applications |
Quantity | Each |
Connector Types | Aseptic |
Unit Size | 1 each |
Description | Transfer Assembly, 1 in. ID Braided Silicone, PendoTECH Pressure Sensor and AseptiQuik L Connectors |
For Use With (Application) | Monoclonal antibody production, upstream and downstream applications |
Quantity | Each |
Connector Types | Aseptic |
Unit Size | 1 each |
Catalog Number | Specifications | Unit Size | Description | Price (ARS) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
SH31245.01 | 1 each | Transfer Assembly, 1 inch ID TPE, PendoTECH Pressure Sensor and AseptiQuik L Connectors | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SH31418.01 | 1 each | Transfer Assembly, 1 in. ID Silicone, PendoTECH Pressure Sensor and AseptiQuik L Connectors | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SH31419.01 | 1 each | Transfer Assembly, 1 in. ID Braided Silicone, PendoTECH Pressure Sensor and AseptiQuik L Connectors | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|