Thermo Scientific™
Transfer Assembly Jumper Tubing, Braided Silicone, Aseptic
Catalog number: SH31391.21
Thermo Scientific™
Transfer Assembly Jumper Tubing, Braided Silicone, Aseptic
Catalog number: SH31391.21
Thermo Scientific™ Standard Fluid Transfer Assemblies with Thermo Scientific™ BioTitan™ Retention Device increases single-use product flexibility. They connect to our standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
Catalog Number
SH31391.21
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID Braided Silicone, AseptiQuik S Connectors
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID Braided Silicone, AseptiQuik S Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik S Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Braided Silicone, AseptiQuik G Connectors
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Braided Silicone, AseptiQuik G Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik G Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Braided Silicone, AseptiQuik G Connectors
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Braided Silicone, AseptiQuik G Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik G Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID Braided Silicone, AseptiQuik G Connectors
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID Braided Silicone, AseptiQuik G Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik G Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Braided Silicone, AseptiQuik S Connectors
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Braided Silicone, AseptiQuik S Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik S Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Braided Silicone, AseptiQuik S Connectors
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Braided Silicone, AseptiQuik S Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik S Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID Braided Silicone, AseptiQuik S Connectors
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID Braided Silicone, AseptiQuik S Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik S Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID Braided Silicone, AseptiQuik G Connectors
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID Braided Silicone, AseptiQuik G Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik G Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 10 ft., 3/8 in. ID Braided Silicone, AseptiQuik G Connectors
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 10 ft., 3/8 in. ID Braided Silicone, AseptiQuik G Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik G Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 15 ft., 3/8 in. ID Braided Silicone, AseptiQuik G Connectors
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 15 ft., 3/8 in. ID Braided Silicone, AseptiQuik G Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik G Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
Showing 10 of 24Show more
Catalog NumberSpecificationsUnit SizeDescriptionPrice (ARS)
SH31391.21Full specifications
1 eachTransfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID Braided Silicone, AseptiQuik S ConnectorsRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID Braided Silicone, AseptiQuik S Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik S Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
SH31389.01Full specifications
1 eachTransfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Braided Silicone, AseptiQuik G ConnectorsRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Braided Silicone, AseptiQuik G Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik G Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
SH31389.02Full specifications
1 eachTransfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Braided Silicone, AseptiQuik G ConnectorsRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Braided Silicone, AseptiQuik G Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik G Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
SH31389.03Full specifications
1 eachTransfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID Braided Silicone, AseptiQuik G ConnectorsRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID Braided Silicone, AseptiQuik G Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik G Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
SH31391.11Full specifications
1 eachTransfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Braided Silicone, AseptiQuik S ConnectorsRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Braided Silicone, AseptiQuik S Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik S Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
SH31391.12Full specifications
1 eachTransfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Braided Silicone, AseptiQuik S ConnectorsRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Braided Silicone, AseptiQuik S Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik S Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
SH31391.13Full specifications
1 eachTransfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID Braided Silicone, AseptiQuik S ConnectorsRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID Braided Silicone, AseptiQuik S Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik S Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
SH31389.11Full specifications
1 eachTransfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID Braided Silicone, AseptiQuik G ConnectorsRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID Braided Silicone, AseptiQuik G Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik G Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
SH31389.12Full specifications
1 eachTransfer Assembly Jumper Tubing, 10 ft., 3/8 in. ID Braided Silicone, AseptiQuik G ConnectorsRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 10 ft., 3/8 in. ID Braided Silicone, AseptiQuik G Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik G Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
SH31389.13Full specifications
1 eachTransfer Assembly Jumper Tubing, 15 ft., 3/8 in. ID Braided Silicone, AseptiQuik G ConnectorsRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 15 ft., 3/8 in. ID Braided Silicone, AseptiQuik G Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptiQuik G Connectors
Tubing MaterialBraided Silicone
Unit Size1 each
Showing 10 of 24Show more
Thermo Scientific™ Standard Fluid Transfer Assemblies with Thermo Scientific™ BioTitan™ Retention Device increases single-use product flexibility. They connect to our standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
• One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
• Manufactured with the BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
• Multiple manufacturing sites ensure security in product supply
• Produced under cGMP conditions in ISO 7 clean rooms to ensure a high level of quality
• Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard BPC products to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common production processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
All Standard Fluid Transfer Assembly products are constructed of capped-end connectors and thermoplastic elastomer (TPE) tubing for use with welders. The Extension Set is available in a variety of tubing sizes and lengths to meet your fluid transfer needs.
These Standard Fluid Transfer Assembly products provide a common set of products that addresses an expansive set of needs. They are manufactured at multiple sites globally to minimize concerns of production interruptions and supply shortages. These products are built to our high quality standards to facilitate the success of your process.
Contents & Storage
Room Temperature