Thermo Scientific™
Transfer Assembly Adapter
Catalog number: SH31429.01
Thermo Scientific™
Transfer Assembly Adapter
Catalog number: SH31429.01
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
Catalog Number
SH31429.01
Unit Size
1 each
Description
Transfer Assembly Adapter, 3/4 in. ID Silicone, AseptiQuik L Connector and MPU Body
Price (ARS)
Full specifications
DescriptionTransfer Assembly Adapter, 3/4 in. ID Silicone, AseptiQuik L Connector and MPU Body
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesNon-aseptic
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Adapter, 3/8 in. ID TPE, MPC body and and AseptiQuik G connector
Price (ARS)
Full specifications
DescriptionTransfer Assembly Adapter, 3/8 in. ID TPE, MPC body and and AseptiQuik G connector
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Adapter, 1 in. ID TPE, AseptiQuik L and G Connectors
Price (ARS)
Full specifications
DescriptionTransfer Assembly Adapter, 1 in. ID TPE, AseptiQuik L and G Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptic
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Adapter, 3/4 to 1/2 in. ID TPE, AseptiQuik L connector and MPX body
Price (ARS)
Full specifications
DescriptionTransfer Assembly Adapter, 3/4 to 1/2 in. ID TPE, AseptiQuik L connector and MPX body
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Adapter, 1 in. ID Silicone, AseptiQuik L and G Connectors
Price (ARS)
Full specifications
DescriptionTransfer Assembly Adapter, 1 in. ID Silicone, AseptiQuik L and G Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptic
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Adapter, 1/2 in. ID TPE, 1 1/2 tri-clamp and MPX insert
Price (ARS)
Full specifications
DescriptionTransfer Assembly Adapter, 1/2 in. ID TPE, 1 1/2 tri-clamp and MPX insert
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Adapter, 1/2 in. ID TPE, MPX body and 3/4 tri-clamp
Price (ARS)
Full specifications
DescriptionTransfer Assembly Adapter, 1/2 in. ID TPE, MPX body and 3/4 tri-clamp
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Adapter, 1/2 in. ID Braided Silicone, 1 1/2 Tri-Clamp and MPX Insert
Price (ARS)
Full specifications
DescriptionTransfer Assembly Adapter, 1/2 in. ID Braided Silicone, 1 1/2 Tri-Clamp and MPX Insert
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesNon-aseptic
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Adapter, 3/4 in. ID TPE, 3/4 tri-clamp and AseptiQuik G connector
Price (ARS)
Full specifications
DescriptionTransfer Assembly Adapter, 3/4 in. ID TPE, 3/4 tri-clamp and AseptiQuik G connector
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Adapter, 3/8 to 1/2 in. ID Braided Silicone, MPC Insert and MPX Insert
Price (ARS)
Full specifications
DescriptionTransfer Assembly Adapter, 3/8 to 1/2 in. ID Braided Silicone, MPC Insert and MPX Insert
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesNon-aseptic
Unit Size1 each
Showing 10 of 45Show more
Catalog NumberSpecificationsUnit SizeDescriptionPrice (ARS)
SH31429.01Full specifications
1 eachTransfer Assembly Adapter, 3/4 in. ID Silicone, AseptiQuik L Connector and MPU BodyRequest A Quote
DescriptionTransfer Assembly Adapter, 3/4 in. ID Silicone, AseptiQuik L Connector and MPU Body
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesNon-aseptic
Unit Size1 each
SH31231.01Full specifications
1 eachTransfer Assembly Adapter, 3/8 in. ID TPE, MPC body and and AseptiQuik G connectorRequest A Quote
DescriptionTransfer Assembly Adapter, 3/8 in. ID TPE, MPC body and and AseptiQuik G connector
Unit Size1 each
SH31375.01Full specifications
1 eachTransfer Assembly Adapter, 1 in. ID TPE, AseptiQuik L and G ConnectorsRequest A Quote
DescriptionTransfer Assembly Adapter, 1 in. ID TPE, AseptiQuik L and G Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptic
Unit Size1 each
SH31232.01Full specifications
1 eachTransfer Assembly Adapter, 3/4 to 1/2 in. ID TPE, AseptiQuik L connector and MPX bodyRequest A Quote
DescriptionTransfer Assembly Adapter, 3/4 to 1/2 in. ID TPE, AseptiQuik L connector and MPX body
Unit Size1 each
SH31428.01Full specifications
1 eachTransfer Assembly Adapter, 1 in. ID Silicone, AseptiQuik L and G ConnectorsRequest A Quote
DescriptionTransfer Assembly Adapter, 1 in. ID Silicone, AseptiQuik L and G Connectors
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesAseptic
Unit Size1 each
SH31233.01Full specifications
1 eachTransfer Assembly Adapter, 1/2 in. ID TPE, 1 1/2 tri-clamp and MPX insertRequest A Quote
DescriptionTransfer Assembly Adapter, 1/2 in. ID TPE, 1 1/2 tri-clamp and MPX insert
Unit Size1 each
SH31234.01Full specifications
1 eachTransfer Assembly Adapter, 1/2 in. ID TPE, MPX body and 3/4 tri-clampRequest A Quote
DescriptionTransfer Assembly Adapter, 1/2 in. ID TPE, MPX body and 3/4 tri-clamp
Unit Size1 each
SH31430.01Full specifications
1 eachTransfer Assembly Adapter, 1/2 in. ID Braided Silicone, 1 1/2 Tri-Clamp and MPX InsertRequest A Quote
DescriptionTransfer Assembly Adapter, 1/2 in. ID Braided Silicone, 1 1/2 Tri-Clamp and MPX Insert
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesNon-aseptic
Unit Size1 each
SH31235.01Full specifications
1 eachTransfer Assembly Adapter, 3/4 in. ID TPE, 3/4 tri-clamp and AseptiQuik G connectorRequest A Quote
DescriptionTransfer Assembly Adapter, 3/4 in. ID TPE, 3/4 tri-clamp and AseptiQuik G connector
Unit Size1 each
SH31435.01Full specifications
1 eachTransfer Assembly Adapter, 3/8 to 1/2 in. ID Braided Silicone, MPC Insert and MPX InsertRequest A Quote
DescriptionTransfer Assembly Adapter, 3/8 to 1/2 in. ID Braided Silicone, MPC Insert and MPX Insert
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesNon-aseptic
Unit Size1 each
Showing 10 of 45Show more
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
• One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
• Manufactured with BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
• Multiple manufacturing sites ensure security in product supply
• Produced under cGMP conditions in ISO 7 clean rooms to ensure a high level of quality
• Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard BPC products to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common production processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
All Standard Fluid Transfer Assembly products are constructed of capped-end connectors and thermoplastic elastomer (TPE) tubing for use with welders. The Extension Set is available in a variety of tubing sizes and lengths to meet your fluid transfer needs.
These Standard Fluid Transfer Assembly products provide a common set of products that addresses an expansive set of needs. They are manufactured at multiple sites globally to minimize concerns of production interruptions and supply shortages. These products are built to our high quality standards to facilitate the success of your process.
Contents & Storage
Room Temperature