DQ Green BSA is a fluorogenic substrate for proteases. A strong fluorescence quenching effect is observed when proteins are heavily labeled with BODIPY dyes. Upon hydrolysis of the DQ Green BSA to single, dye-labeled peptides by proteases, this quenching is relieved, producing brightly fluorescent products.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection Method
Fluorescence
Form
Solid
Product Line
DQ™
Quantity
5 x 1 mg
Substrate
Protease Substrate
Substrate Properties
Protein-Based Substrate
Shipping Condition
Room Temperature
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.