Copper(II) nitrate trihydrate, 99%, for analysis
Copper(II) nitrate trihydrate, 99%, for analysis
Copper(II) nitrate trihydrate, 99%, for analysis
Copper(II) nitrate trihydrate, 99%, for analysis
Thermo Scientific Chemicals

Copper(II) nitrate trihydrate, 99%, for analysis

CAS: 10031-43-3 | CuN2O6·3H2O | 241.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 207685000
Price (EUR)
61,71
Online Exclusive
74,40
Save 12,69 (17%)
Each
In stock
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
61,71
Online Exclusive
74,40
Save 12,69 (17%)
Each
Add to cart
Chemical Identifiers
CAS10031-43-3
IUPAC Namecopper(2+) dinitrate
Molecular FormulaCuN2O6
InChI KeyXTVVROIMIGLXTD-UHFFFAOYSA-N
SMILES[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Iodometric98.5 to 104.0 %
Sulfate (SO4)=<50 ppm
Zinc (Zn)=<50 ppm
Appearance (Color)Blue
Appearance (Form)Powder or crystals and/or chunks
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470793Certificate of AnalysisMay 17, 2025207680025, 207681000, 207685000
A0465183Certificate of AnalysisAug 15, 2024207680025, 207681000, 207685000
A0460173Certificate of AnalysisFeb 19, 2024207680025, 207681000, 207685000
A0454704Certificate of AnalysisAug 25, 2023207680025, 207681000, 207685000
A0401275Certificate of AnalysisDec 15, 2022207680025, 207681000, 207685000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Oxidizing solids
Category 2
Skin Corrosion/Irritation
Category 1
Serious Eye Damage/Eye Irritation
Category 1
Acute aquatic toxicity
Category 1
Chronic aquatic toxicity
Category 2
Label Elements
Signal Word

Danger

Hazard Statements

H272 - May intensify fire; oxidizer

H314 - Causes severe skin burns and eye damage

H400 - Very toxic to aquatic life

H411 - Toxic to aquatic life with long lasting effects

Precautionary Statements

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

P220 - Keep away from clothing and other combustible materials

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

P371 + P380 + P375 - In case of major fire and large quantities: Evacuate area. Fight fire remotely due to the risk of explosion

Share catalog number, name or link

1x1 image pixel for data collection