Pyrroloquinoline quinone
Pyrroloquinoline quinone
Thermo Scientific Chemicals

Pyrroloquinoline quinone

CAS: 72909-34-3 | C14H6N2O8, C14H6N2O8 | 330.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 467010010
Price (EUR)
559,30
Online Exclusive
674,00
Save 114,70 (17%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
1 g
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
559,30
Online Exclusive
674,00
Save 114,70 (17%)
Each
Add to cart
Chemical Identifiers
CAS72909-34-3
IUPAC Name4,5-dioxo-1H,4H,5H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid
Molecular FormulaC14H6N2O8
InChI KeyMMXZSJMASHPLLR-UHFFFAOYSA-N
SMILESOC(=O)C1=CC2=C(N1)C1=C(C=C(N=C1C(=O)C2=O)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)powder
Infrared spectrumConforms
HPLC>=95.0 %
Appearance (Color)Dark orange to red to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0464415Certificate of AnalysisSep 06, 2024467012500, 467010010
A0462885Certificate of AnalysisJun 10, 2024467012500, 467010010
A0447588Certificate of AnalysisNov 30, 2022467012500, 467010010
A0443623Certificate of AnalysisJul 27, 2022467012500, 467010010
A0419185Certificate of AnalysisJul 08, 2020467012500, 467010010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H302 - Harmful if swallowed

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P312 - Call a POISON CENTER or doctor if you feel unwell

Share catalog number, name or link

1x1 image pixel for data collection