3-Methoxy-2-nitrobenzaldehyde, 97%
3-Methoxy-2-nitrobenzaldehyde, 97%
3-Methoxy-2-nitrobenzaldehyde, 97%
3-Methoxy-2-nitrobenzaldehyde, 97%
Thermo Scientific Chemicals

3-Methoxy-2-nitrobenzaldehyde, 97%

CAS: 53055-05-3 | C8H7NO4 | 181.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 172670250
Price (EUR)
515,95
Online Exclusive
622,00
Save 106,05 (17%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
25 g
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
515,95
Online Exclusive
622,00
Save 106,05 (17%)
Each
Add to cart
Chemical Identifiers
CAS53055-05-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point97°C to 103°C
Infrared spectrumConforms
Appearance (Color)White to yellow to beige
GC>=96.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
344268Certificate of AnalysisDec 12, 2022172670010, 172670050, 172670250
343724Certificate of AnalysisMar 19, 2021172670010, 172670050, 172670250
2 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

Precautionary Statements

P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

Share catalog number, name or link