Éter fenil, 99 %, Thermo Scientific Chemicals
Éter fenil, 99 %, Thermo Scientific Chemicals
Éter fenil, 99 %, Thermo Scientific Chemicals
Éter fenil, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Éter fenil, 99 %, Thermo Scientific Chemicals

CAS: 101-84-8 | C12H10O | 170.21 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 kg
100 g
500 g
2.5 kg
10 kg
Envase:
Ver todo
Tambor de plástico
Número de catálogo 130600100
Precio (EUR)
213,35
Online Exclusive
257,00
Ahorro 43,65 (17%)
Each
-
Añadir al carro de la compra
Cantidad:
10 kg
Envase:
Tambor de plástico
Pedido a granel o personalizado
Precio (EUR)
213,35
Online Exclusive
257,00
Ahorro 43,65 (17%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS101-84-8
IUPAC Namephenoxybenzene
Molecular FormulaC12H10O
InChI KeyUSIUVYZYUHIAEV-UHFFFAOYSA-N
SMILESO(C1=CC=CC=C1)C1=CC=CC=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Liquid after melting
Appearance (Color)Clear light yellow
Infrared spectrumConforms
Refractive index1.5795 to 1.5815 (20°C, 589 nm)
GC>=98.5 %
Ask our AI about this Product

Este producto de marca Thermo Scientific Chemicals formó parte originalmente de la cartera de productos Acros Organics. Alguna información sobre documentos y etiquetas puede referirse a la marca original. El producto original Acros Organics/código de artículo o referencia SKU no ha cambiado como parte de la transición de marca a Thermo Scientific Chemicals.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documentos y descargas

Certificados

N.º de loteCertificate TypeDateCatalog Number(s)
A0473393Certificate of Analysis26 jun 2025130605000, 130601000, 130600010, 130600100, 130600025
A0471448Certificate of Analysis17 may 2025130605000, 130601000, 130600010, 130600100, 130600025
A0448250Certificate of Analysis31 ene 2025130605000, 130601000, 130600010, 130600100, 130600025
A0467070Certificate of Analysis31 ene 2025130605000, 130601000, 130600010, 130600100, 130600025
A0448250Certificate of Analysis25 dic 2023130605000, 130601000, 130600010, 130600100, 130600025
Se muestran 5 resultados, busque arriba un certificado específico

Hojas de datos de seguridad

Seguridad y manipulación

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Serious Eye Damage/Eye Irritation
Category 2
Skin Sensitization
Category 1
Skin Sensitization
Sub-category 1B
Reproductive Toxicity
Category 1B
Acute aquatic toxicity
Category 1
Chronic aquatic toxicity
Category 3
Label Elements
Signal Word

Danger

Hazard Statements

H317 - May cause an allergic skin reaction

H319 - Causes serious eye irritation

H360FD - May damage fertility. May damage the unborn child

H400 - Very toxic to aquatic life

H412 - Harmful to aquatic life with long lasting effects

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P308 + P313 - IF exposed or concerned: Get medical advice/attention

Additional EU labelling

Restricted to professional users

Compartir número de catálogo, nombre o enlace.

1x1 image pixel for data collection