Ácido 3-(bromometil)fenilborónico, 97 %, Thermo Scientific Chemicals
Ácido 3-(bromometil)fenilborónico, 97 %, Thermo Scientific Chemicals
Ácido 3-(bromometil)fenilborónico, 97 %, Thermo Scientific Chemicals
Ácido 3-(bromometil)fenilborónico, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido 3-(bromometil)fenilborónico, 97 %, Thermo Scientific Chemicals

CAS: 51323-43-4 | C7H8BBrO2 | 214.85 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Envase:
Frasco de vidrio
Número de catálogo 430600010
Precio (EUR)
31,45
Exklusiv online
37,90
Ahorro 6,45 (17%)
Each
-
Añadir al carro de la compra
Cantidad:
1 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
31,45
Exklusiv online
37,90
Ahorro 6,45 (17%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS51323-43-4
IUPAC Name[3-(bromomethyl)phenyl]boronic acid
Molecular FormulaC7H8BBrO2
InChI KeyATRFDLFMCLYROQ-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC(CBr)=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Powder or crystalline powder
Appearance (Color)White to tan
Infrared spectrumConforms
HPLC>=96.0 %
Ask our AI about this Product
Este producto de marca Thermo Scientific Chemicals formó parte originalmente de la cartera de productos Acros Organics.Alguna información sobre documentos y etiquetas puede referirse a la marca original.El producto original de Acros Organics / código de artículo o referencia SKU no ha cambiado como parte de la transición de marca a Thermo Scientific Chemicals.
RUO - Para uso exclusivo en investigación
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documentos y descargas

Certificados

N.º de loteCertificate TypeDateCatalog Number(s)
345149Certificate of Analysis25 ago 2023430600010, 430600050
345047Certificate of Analysis08 feb 2023430600010, 430600050
342355Certificate of Analysis29 nov 2021430600010, 430600050
Se muestran 3 resultados, busque arriba un certificado específico

Hojas de datos de seguridad

Seguridad y manipulación

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P312 - Call a POISON CENTER or doctor if you feel unwell

Compartir número de catálogo, nombre o enlace.

1x1 image pixel for data collection