1x1 image pixel for data collection
Valsartán
Thermo Scientific Chemicals

Valsartán

CAS: 137862-53-4 | C24H29N5O3 | 435.53 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
1 g
Número de catálogo 465560050
Precio (EUR)
96,05
Online Exclusive
116,00
Ahorro 19,95 (17%)
Each
Disponibilidad estimada  Pendientes  
Añadir al carro de la compra
Cantidad:
5 g
Recurring order eligible. Learn more »
Pedido a granel o personalizado
Precio (EUR)
96,05
Online Exclusive
116,00
Ahorro 19,95 (17%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS137862-53-4
IUPAC Name(2S)-3-methyl-2-(N-{[2'-(2H-1,2,3,4-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl]methyl}pentanamido)butanoic acid
Molecular FormulaC24H29N5O3
InChI KeyACWBQPMHZXGDFX-QFIPXVFZSA-N
SMILESCCCCC(=O)N(CC1=CC=C(C=C1)C1=CC=CC=C1C1=NNN=N1)[C@@H](C(C)C)C(O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Specific optical rotation-55° to -70° (20°C, 589 nm) (1% in CH3OH)
Water=<2 %
Appearance (Color)White to almost white to light beige
Appearance (Form)Powder
Infrared spectrumConforms
Ver más
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documentos y descargas

Certificados

N.º de loteCertificate TypeDateCatalog Number(s)
A0472607Certificate of Analysis20 may 2025465560010, 465560050
A0430578Certificate of Analysis10 jun 2024465560010, 465560050
A0461290Certificate of Analysis18 abr 2024465560010, 465560050
A0458990Certificate of Analysis21 feb 2024465560010, 465560050
A0443972Certificate of Analysis24 ene 2024465560010, 465560050
Se muestran 5 resultados, busque arriba un certificado específico

Hojas de datos de seguridad

Seguridad y manipulación

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Reproductive Toxicity
Category 2
Label Elements
Signal Word

Warning

Hazard Statements

H361 - Suspected of damaging fertility or the unborn child

Precautionary Statements

P201 - Obtain special instructions before use

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P308 + P313 - IF exposed or concerned: Get medical advice/attention

Compartir número de catálogo, nombre o enlace.

1x1 image pixel for data collection