Sodium hypochlorite, 4-6%
Sodium hypochlorite, 4-6%
Sodium hypochlorite, 4-6%
Sodium hypochlorite, 4-6%
Thermo Scientific Chemicals

Sodium hypochlorite, 4-6%

CAS: 7681-52-9 | ClNaO | 74.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 419550010
Price (EUR)
141,95
キャンペーン価格
167,00
Save 25,05 (15%)
Each
-
Add to cart
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
141,95
キャンペーン価格
167,00
Save 25,05 (15%)
Each
Add to cart
Chemical Identifiers
CAS7681-52-9
IUPAC Namesodium hypochlorite
Molecular FormulaClNaO
InChI KeySUKJFIGYRHOWBL-UHFFFAOYSA-N
SMILES[Na+].[O-]Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Appearance (Form)Solution
Titration Iodometric4.0 to 6.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470093Certificate of AnalysisMay 17, 2025419550100, 419552500, 419550025, 419550250, 419550010
A0466651Certificate of AnalysisMay 17, 2025419550100, 419552500, 419550025, 419550250, 419550010
A0462548Certificate of AnalysisJan 31, 2025419550100, 419552500, 419550025, 419550250, 419550010
A0466651Certificate of AnalysisOct 08, 2024419550100, 419552500, 419550025, 419550250, 419550010
A0462548Certificate of AnalysisMay 06, 2024419550100, 419552500, 419550025, 419550250, 419550010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Substances/mixtures corrosive to metal
Category 1
Skin Corrosion/Irritation
Category 1
Serious Eye Damage/Eye Irritation
Category 1
Acute aquatic toxicity
Category 1
Chronic aquatic toxicity
Category 2
Label Elements
Signal Word

Danger

Hazard Statements

H314 - Causes severe skin burns and eye damage

H400 - Very toxic to aquatic life

H411 - Toxic to aquatic life with long lasting effects

Physical hazards

H290 - May be corrosive to metals

EU Specific Hazard Statements

EUH031 - Contact with acids liberates toxic gas

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

Share catalog number, name or link

1x1 image pixel for data collection