Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CCCACTGGCTGGGAAGACACAGAGG[A/T]GCTACGGCTGGTGCGGCCACCCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300408 MIM: 300281 MIM: 300713 | ||||||||||||||||||||
Literature Links: |
GRIPAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRIPAP1 - GRIP1 associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCND1 - potassium voltage-gated channel subfamily D member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004979.5 | 1858 | Missense Mutation | ACC,TCC | T,S 516 | NP_004970.3 | |
XM_011543910.2 | 1858 | Missense Mutation | ACC,TCC | T,S 516 | XP_011542212.1 | |
XM_017029508.1 | 1858 | Missense Mutation | ACC,TCC | T,S 427 | XP_016884997.1 | |
XM_017029509.1 | 1858 | Missense Mutation | ACC,TCC | T,S 427 | XP_016884998.1 | |
XM_017029510.1 | 1858 | Intron | XP_016884999.1 |
OTUD5 - OTU deubiquitinase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |