Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | mmu-miR-92a-1* |
---|---|
miRBase Accession Number: | MI0000719 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AGGUUGGGAUUUGUCGCAAUGCU |
Species: | Mouse, Rat |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000013, mir-25 |
Mature miRNA Sequence: | AGGUUGGGAUUUGUCGCAAUGCU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-92a-1-5p | MIMAT0017066 |
Rat | rno-miR-92a-1-5p | MIMAT0017107 |