Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Amplicon Length: | 132 |
Chromosome Location: | Chr. 15 - 100180852 - 100180852 [-] on 100180852 Build Rnor_6.0 |
Species: | Rat |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-19b-1-5p | MIMAT0017096 |
Rat | rno-miR-19b-3p | MIMAT0000788 |
Rat | rno-miR-92a-3p | MIMAT0000816 |
Rat | rno-miR-92a-1-5p | MIMAT0017107 |