Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-363-3p |
---|---|
Stem-loop Accession Number: | MI0000764 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AAUUGCACGGUAUCCAUCUGUA |
Species: | Human, Mouse, Callithrix jacchus, Cyprinus carpio, Dasypus novemcinctus, Guinea pig, Horse, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Tupaia chinensis, Xenopus laevis, Zebrafish |
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000138, mir-363 |
Mature miRNA Sequence: | AAUUGCACGGUAUCCAUCUGUA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-363-3p | MIMAT0000707 |
Mouse | mmu-miR-363-3p | MIMAT0000708 |
Callithrix jacchus | cja-miR-363 | MIMAT0039359 |
Cyprinus carpio | ccr-miR-363 | MIMAT0026293 |
Dasypus novemcinctus | dno-miR-363-3p | MIMAT0047834 |
Guinea pig | cpo-miR-363-3p | MIMAT0047193 |
View More Rows | ||
Horse | eca-miR-363 | MIMAT0013212 |
Pongo pygmaeus | ppy-miR-363 | MIMAT0015852 |
Pteropus alecto | pal-miR-363-3p | MIMAT0039877 |
Rabbit | ocu-miR-363-3p | MIMAT0048398 |
Rhesus monkey | mml-miR-363-3p | MIMAT0006290 |
Tupaia chinensis | tch-miR-363-3p | MIMAT0036586 |
Xenopus laevis | xla-miR-363-3p | MIMAT0046589 |
Zebrafish | dre-miR-363-3p | MIMAT0001874 |