Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Amplicon Length: | 149 |
Chromosome Location: | Chr. X - 134169188 - 134169188 [-] on 134169188 Build GRCh38 |
Species: | Human |
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-19b-3p | MIMAT0000074 | |
Human | hsa-miR-19b-2-5p | MIMAT0004492 | |
Human | hsa-miR-20b-3p | MIMAT0004752 | |
Human | hsa-miR-20b-5p | MIMAT0001413 | |
Human | hsa-miR-363-3p | MIMAT0000707 | |
Human | hsa-miR-363-5p | MIMAT0003385 | |
View More Rows | |||
Human | hsa-miR-92a-2-5p | MIMAT0004508 | |
Human | hsa-miR-92a-3p | MIMAT0000092 |