Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-18a-5p |
---|---|
Stem-loop Accession Number: | MI0000072 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UAAGGUGCAUCUAGUGCAGAUAG |
Species: | Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Goat, Guinea pig, Horse, Microcebus murinus, Ophiophagus hannah, Oreochromis niloticus, Oryzias latipes, Otolemur garnettii, Petromyzon marinus, Python bivittatus, Rabbit, Xenopus tropicalis |
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | UAAGGUGCAUCUAGUGCAGAUAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-18a-5p | MIMAT0000072 |
Mouse | mmu-miR-18a-5p | MIMAT0000528 |
Rat | rno-miR-18a-5p | MIMAT0000787 |
Alligator mississippiensis | ami-miR-18-5p | MIMAT0038049 |
Anolis carolinensis | aca-miR-18a-5p | MIMAT0021817 |
Cricetulus griseus | cgr-miR-18a-5p | MIMAT0023817 |
View More Rows | ||
Cyprinus carpio | ccr-miR-18a | MIMAT0026246 |
Dasypus novemcinctus | dno-miR-18a-5p | MIMAT0047511 |
Goat | chi-miR-18a-5p | MIMAT0036015 |
Guinea pig | cpo-miR-18a-5p | MIMAT0046859 |
Horse | eca-miR-18a | MIMAT0013085 |
Microcebus murinus | mmr-miR-18 | MIMAT0049197 |
Ophiophagus hannah | oha-miR-18a-5p | MIMAT0036768 |
Oreochromis niloticus | oni-miR-18a-2 | MIMAT0043044 |
Oryzias latipes | ola-miR-18 | MIMAT0022532 |
Otolemur garnettii | oga-miR-18 | MIMAT0049647 |
Petromyzon marinus | pma-miR-18a-5p | MIMAT0019387 |
Python bivittatus | pbv-miR-18a-5p | MIMAT0038840 |
Rabbit | ocu-miR-18a-5p | MIMAT0048111 |
Xenopus tropicalis | xtr-miR-18a-5p | MIMAT0003652 |