Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Amplicon Length: | 99 |
Chromosome Location: | Chr. 15 - 100179709 - 100179709 [-] on 100179709 Build Rnor_6.0 |
Species: | Rat |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-17-1-3p | MIMAT0004710 |
Rat | rno-miR-17-5p | MIMAT0000786 |
Rat | rno-miR-18a-5p | MIMAT0000787 |
Rat | rno-miR-18a-3p | MIMAT0017095 |
Rat | rno-miR-19a-3p | MIMAT0000789 |
Rat | rno-miR-19a-5p | MIMAT0017098 |