Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-363-5p |
---|---|
Stem-loop Accession Number: | MI0000764 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CGGGUGGAUCACGAUGCAAUUU |
Species: | Human, Rat, Goat, Ophiophagus hannah, Ornithorhynchus anatinus, Xenopus laevis, Xenopus tropicalis |
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000138, mir-363 |
Mature miRNA Sequence: | CGGGUGGAUCACGAUGCAAUUU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-363-5p | MIMAT0003385 |
Rat | rno-miR-363-5p | MIMAT0003209 |
Goat | chi-miR-363-5p | MIMAT0036175 |
Ophiophagus hannah | oha-miR-363-5p | MIMAT0036912 |
Ornithorhynchus anatinus | oan-miR-363-5p | MIMAT0006859 |
Xenopus laevis | xla-miR-363 | MIMAT0011147 |
View More Rows | ||
Xenopus tropicalis | xtr-miR-363-5p | MIMAT0003701 |