Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-19b-2-5p |
---|---|
miRBase Accession Number: | MI0000075 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AGUUUUGCAGGUUUGCAUUUCA |
Species: | Human |
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000011, mir-19 |
Mature miRNA Sequence: | AGUUUUGCAGGUUUGCAUUUCA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-19b-2-5p | MIMAT0004492 |