Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | rno-miR-20b-3p |
---|---|
miRBase Accession Number: | MI0003554 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | ACUGCAGUGUGAGCACUUCUGG |
Species: | Rat |
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | ACUGCAGUGUGAGCACUUCUGG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-20b-3p | MIMAT0003212 |