Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Amplicon Length: | 82 |
Chromosome Location: | Chr. X - 140117746 - 140117746 [-] on 140117746 Build Rnor_6.0 |
Species: | Rat |
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Rat | rno-miR-17-5p | MIMAT0000786 | |
Rat | rno-miR-17-2-3p | MIMAT0017295 | |
Rat | rno-miR-19b-2-5p | MIMAT0017097 | |
Rat | rno-miR-19b-3p | MIMAT0000788 | |
Rat | rno-miR-20b-3p | MIMAT0003212 | |
Rat | rno-miR-20b-5p | MIMAT0003211 | |
View More Rows | |||
Rat | rno-miR-92a-2-5p | MIMAT0017108 | |
Rat | rno-miR-92a-3p | MIMAT0000816 |