Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-20b-3p |
---|---|
miRBase Accession Number: | MI0001519 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | ACUGUAGUAUGGGCACUUCCAG |
Species: | Human |
Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | ACUGUAGUAUGGGCACUUCCAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-20b-3p | MIMAT0004752 |