Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | rno-miR-92a-2-5p |
---|---|
miRBase Accession Number: | MI0000879 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AGGUGGGGAUUAGUGCCAUUAC |
Species: | Rat |
Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000013, mir-25 |
Mature miRNA Sequence: | AGGUGGGGAUUAGUGCCAUUAC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-92a-2-5p | MIMAT0017108 |